SY3H3D677513 Synthonix Corporation

3-Bromo-4-(trifluoromethyl)aniline

  • MDL number MFCD12828280
  • Cas Number : 172215-91-7
  • Molecular Weight : 240.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown to Brown to Red Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44900.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44900.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart