SY3H3D677531 Synthonix Corporation

2-Bromo-5-(trifluoromethoxy)aniline

  • MDL number MFCD08458013
  • Cas Number : 887267-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45034.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45034.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart