SY3H3D677547 Synthonix Corporation

4-Bromo-3-hydroxybenzoic acid

  • MDL number MFCD08056371
  • Cas Number : 14348-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45113.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45113.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart