SY3H3D677561 Synthonix Corporation

2-Bromo-6-fluorotoluene

  • MDL number MFCD00051438
  • Cas Number : 1422-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45164.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45164.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart