SY3H3D677569 Synthonix Corporation

1-Bromo-2-iodo-3-methylbenzene

  • MDL number MFCD12547770
  • Cas Number : 869500-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45172.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45172.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart