SY3H3D67756A Synthonix Corporation

1-Bromo-4-iodo-2-methylbenzene

  • MDL number MFCD00070749
  • Cas Number : 202865-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Slightly pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45173.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45173.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart