SY3H3D677572 Synthonix Corporation

Benzo[d]isoxazol-3-amine

  • MDL number MFCD03407353
  • Cas Number : 36216-80-5
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45214.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45214.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart