SY3H3D677578 Synthonix Corporation

1-Bromo-3-methylimidazo[1,5-a]pyrazine

  • MDL number MFCD09999201
  • Cas Number : 56481-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow to Red Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45274.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45274.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart