SY3H3D677580 Synthonix Corporation

6-Bromo-5-methyl-1H-imidazo[4,5-b]pyridine

  • MDL number MFCD00225747
  • Cas Number : 28279-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45282.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45282.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart