SY3H3D677588 Synthonix Corporation

2-Bromo-5-(difluoromethyl)-4-methylpyridine

  • MDL number MFCD25477783
  • Cas Number : 1805019-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45318.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45318.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart