SY3H3D67758C Synthonix Corporation

3-Bromo-5-methylpicolinic acid

  • MDL number MFCD17214206
  • Cas Number : 1211515-68-2
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to Yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45334.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45334.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart