SY3H3D67759C Synthonix Corporation

4-Bromo-2-nitrobenzylalcohol

  • MDL number MFCD07779493
  • Cas Number : 22996-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45405.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/45405.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart