SY3H3D6775A6 Synthonix Corporation

6-Bromo-5-methoxypicolinic acid

  • MDL number MFCD06655957
  • Cas Number : 54232-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form WHITE SOLID
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45415.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45415.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart