SY3H3D6775A7 Synthonix Corporation

1-Bromo-3-methoxy-2-nitrobenzene

  • MDL number MFCD11101044
  • Cas Number : 500298-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Slightly pale yellow - Pale yellow green - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45416.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45416.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart