SY3H3D6775BF Synthonix Corporation

5-Borono-2-chlorobenzoic acid

  • MDL number MFCD06801689
  • Cas Number : 913835-32-2
  • Molecular Weight : 200.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45533.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45533.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart