SY3H3D6775C2 Synthonix Corporation

4-Borono-3-fluorobenzoic acid

  • MDL number MFCD08458473
  • Cas Number : 851335-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45538.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45538.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart