SY3H3D6775C9 Synthonix Corporation

(2-Bromo-5-chlorophenyl)methanol

  • MDL number MFCD09261147
  • Cas Number : 60666-70-8
  • Molecular Weight : 221.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white to gray powder or crysttals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45558.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45558.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart