SY3H3D6775CE Synthonix Corporation

2-Bromo-1-chloro-4-methoxybenzene

  • MDL number MFCD00070739
  • Cas Number : 2732-80-1
  • Molecular Weight : 221.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45563.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45563.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart