SY3H3D6779C2 Synthonix Corporation

1-Bromo-3-fluoro-5-methoxybenzene

  • MDL number MFCD01861129
  • Cas Number : 29578-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Colorless to Pale-yellow Solid or Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45574.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45574.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart