SY3H3D6779C4 Synthonix Corporation

2-Bromo-1-fluoro-3-methoxybenzene

  • MDL number MFCD03788540
  • Cas Number : 446-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to pale-yellow to brown solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45576.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45576.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart