SY3H3D677A01 Synthonix Corporation

6-Bromo-2-chloro-3-methoxy-4-methylpyridine

  • MDL number MFCD22628324
  • Cas Number : 1403764-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45876.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45876.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart