SY3H3D677A05 Synthonix Corporation

(5-Bromo-2-fluoro-3-methylphenyl)boronic acid

  • MDL number MFCD09800868
  • Cas Number : 957120-61-5
  • Molecular Weight : 232.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45893.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45893.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart