SY3H3D677A12 Synthonix Corporation

(6-Bromo-3-fluoro-2-methoxyphenyl)boronic acid

  • MDL number MFCD09475867
  • Cas Number : 957035-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45929.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45929.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart