SY3H3D677A38 Synthonix Corporation

4-Bromo-N-methylbenzenesulfonamide

  • MDL number MFCD00993162
  • Cas Number : 703-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46133.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46133.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart