SY3H3D677A3B Synthonix Corporation

2-Bromo-3-methoxyaniline

  • MDL number MFCD07787450
  • Cas Number : 112970-44-2
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Yellow to Brown or Gray Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/46150.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46150.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart