SY3H3D677A6B Synthonix Corporation

2-Bromo-4,5,6,7-tetrahydrobenzo[d]thiazol-6-amine

  • MDL number MFCD13195381
  • Cas Number : 1379295-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46297.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46297.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart