SY3H3D677A6C Synthonix Corporation

5-Bromo-4,6-dimethoxy-2-methylpyrimidine

  • MDL number MFCD26517008
  • Cas Number : 4319-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to Off-white to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46312.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46312.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart