SY3H3D677A78 Synthonix Corporation

1-Butyl-3-methylimidazolium iodide

  • MDL number MFCD08276307
  • Cas Number : 65039-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Yellow - Deep reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46621.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46621.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart