SY3H3D677A7D Synthonix Corporation

1-(5-Bromopyrimidin-2-yl)-3-pyrrolidinol

  • MDL number MFCD08460364
  • Cas Number : 914347-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46713.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46713.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart