SY3H3D677A91 Synthonix Corporation

1-(3-Bromophenyl)ethylamine Hydrochloride

  • MDL number MFCD26960538
  • Cas Number : 90151-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46739.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46739.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart