SY3H3D677A9C Synthonix Corporation

4-Bromo-2-isopropoxypyridine

  • MDL number MFCD11869614
  • Cas Number : 1142194-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46785.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46785.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart