SY3H3D677AA6 Synthonix Corporation

5-Bromo-N-isopropylthiophene-2-carboxamide

  • MDL number MFCD08593344
  • Cas Number : 908494-87-1
  • Molecular Weight : 248.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to light-yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46800.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46800.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart