SY3H3D677AA8 Synthonix Corporation

Bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 120-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46835.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46835.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart