SY3H3D677AAD Synthonix Corporation

1-Butyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD21668277
  • Cas Number : 1426958-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46865.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46865.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart