SY3H3D677AAF Synthonix Corporation

tert-Butyl (2-bromothiazol-4-yl)carbamate

  • MDL number MFCD12032212
  • Cas Number : 1245647-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46867.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46867.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart