SY3H3D677AC3 Synthonix Corporation

N-tert-Butyl-2-thiophenesulfonamide

  • MDL number MFCD02047252
  • Cas Number : 100342-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Green - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47062.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47062.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart