SY3H3D677AC9 Synthonix Corporation

Bicyclo[2.2.2]octan-2-ol

  • MDL number MFCD00218769
  • Cas Number : 18684-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47081.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47081.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart