SY3H3D677AD7 Synthonix Corporation

(R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid hydrate

  • MDL number MFCD26792600
  • Cas Number : 350230-37-4
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47139.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47139.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart