SY3H3D677ADD Synthonix Corporation

tert-Butyl (1-bromopropan-2-yl)carbamate

  • MDL number MFCD24167884
  • Cas Number : 1391026-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47154.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47154.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart