SY3H3D677AE4 Synthonix Corporation

tert-Butyl (2,3-dihydroxypropyl)carbamate

  • MDL number MFCD01321265
  • Cas Number : 137618-48-5
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form powder, crystalline Colour: white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47186.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47186.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart