SY3H3D677AED Synthonix Corporation

tert-Butyl (3-amino-2-hydroxypropyl)carbamate

  • MDL number MFCD04118107
  • Cas Number : 144912-84-5
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47203.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47203.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart