SY3H3D677AF4 Synthonix Corporation

2-((tert-Butyldimethylsilyl)oxy)ethanol

  • MDL number No MDL found
  • Cas Number : 102229-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47219.html

Safety Information

signalword Warning
pcodes P210, P261
MSDS https://synthonix.com/47219.html
pictograms GHS02
hstatements H225, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart