SY3H3D677B02 Synthonix Corporation

4-Bromo-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD06659466
  • Cas Number : 191165-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Lump / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47270.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47270.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart