SY3H3D677B10 Synthonix Corporation

8-Bromo-2-fluoro-1,5-naphthyridine

  • MDL number No MDL found
  • Cas Number : 1432322-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47328.html

Safety Information

MSDS https://synthonix.com/47328.html
Availability details for
Quantity
  • Availability
  • Add to Cart