SY3H3D677B1A Synthonix Corporation

2-(5-Bromo-2-chloro-4-fluorophenyl)acetonitrile

  • MDL number MFCD23166759
  • Cas Number : 1426290-08-5
  • Molecular Weight : 248.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47376.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47376.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart