SY3H3D677B1C Synthonix Corporation

4-Bromoisatin

  • MDL number MFCD03618555
  • Cas Number : 20780-72-7
  • Molecular Weight : 226.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47382.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47382.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart