SY3H3D677B25 Synthonix Corporation

5-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione

  • MDL number MFCD15526667
  • Cas Number : 77603-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47412.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47412.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart