SY3H3D677B2F Synthonix Corporation

2-Bromo-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD06208253
  • Cas Number : 1483-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47462.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47462.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart