SY3H3D677B46 Synthonix Corporation

2-Bromobenzo[b]thiophene

  • MDL number MFCD08435846
  • Cas Number : 5394-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47534.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47534.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart