SY3H3D677B4B Synthonix Corporation

2-Bromoquinazoline

  • MDL number MFCD19686152
  • Cas Number : 1316275-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47544.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47544.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart